1-Methylpyrrolidin-3-amine 95%

Supplier: Apollo Scientific

Synonyms: 3-Amino-1-methylpyrrolidine

OR308139-1G OR308139-5G OR308139-25G
APOSOR308139-1GEA 178 EUR
APOSOR308139-1G APOSOR308139-5G APOSOR308139-25G
1-Methylpyrrolidin-3-amine 95%
1-Methylpyrrolidin-3-amine
Formula: C₅H₁₂N₂
MW: 100,16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09971801
CAS Number: 13220-27-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR