(S)-4-Boc-(3-hydroxymethyl)morpholine
Supplier: Apollo Scientific
Synonyms:
tert-Butyl (3S)-3-(hydroxymethyl)morpholine-4-carboxylate
OR308157-1G
OR308157-5G
APOSOR308157-1GEA
115
EUR
APOSOR308157-1G
APOSOR308157-5G
(S)-4-Boc-(3-hydroxymethyl)morpholine
(S)-4-Boc-(3-hydroxymethyl)morpholine
Formula:
C₁₀H₁₉NO₄ MW: 217,27 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06799478 CAS Number: 714971-28-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...