Azetidine-2-carboxylic acid 98%

Supplier: Apollo Scientific

Synonyms: azetidine-2-carboxylic acid

OR309022-5G
APOSOR309022-5GEA 1150 EUR
APOSOR309022-5G
Azetidine-2-carboxylic acid 98%
Azetidine-2-carboxylic acid
Formula: C₄H₇NO₂
MW: 101,11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00066660
CAS Number: 2517-04-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR