Azetidine-3-carbonitrile hydrochloride 95%
Supplier: Apollo Scientific
|
Danger
|
OR309045-1G
OR309045-5G
APOSOR309045-1GEA
27.7
EUR
APOSOR309045-1G
APOSOR309045-5G
Azetidine-3-carbonitrile hydrochloride 95%
Azetidine-3-carbonitrile hydrochloride
Formula:
C₄H₇ClN₂ MW: 118,57 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09749828 CAS Number: 345954-83-8 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...