2-Methyl-N-(3-oxetanylidene)propane-2-sulphinamide
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2-Methyl-N-(oxetan-3-ylidene)propane-2-sulphinamide
OR309075-1G
OR309075-250MG
APOSOR309075-1GEA
109
EUR
APOSOR309075-1G
APOSOR309075-250MG
2-Methyl-N-(3-oxetanylidene)propane-2-sulphinamide
2-Methyl-N-(3-oxetanylidene)propane-2-sulphinamide
Formula:
C₇H₁₃NO₂S MW: 175,25 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD14702522 CAS Number: 1158098-73-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...