Pyrrolidin-3-ylmethanol
Supplier: Apollo Scientific
|
Warning
|
OR309156-5G
APOSOR309156-5GEA
140
EUR
APOSOR309156-5G
Pyrrolidin-3-ylmethanol
Pyrrolidin-3-ylmethanol
Formula:
C5H11NO MW: 101,15 g/mol Boiling Pt: 165…171 °C Density: 0,979 g/cm³ (25 °C) Flash Pt: 86 °C Storage Temperature: Ambient |
MDL Number:
MFCD07773080 CAS Number: 5082-74-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...