3-(Methylamino)-1-propanol
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
3-(Methylamino)propan-1-ol
OR309167-25G
APOSOR309167-25GEA
119
EUR
APOSOR309167-25G
3-(Methylamino)-1-propanol
3-(Methylamino)-1-propanol
Formula:
C₄H₁₁NO MW: 89,14 g/mol Boiling Pt: 169 °C Density: 0,98 g/cm³ (25 °C) Flash Pt: 82 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD06797456 CAS Number: 42055-15-2 UN: 2735 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...