N-Benzyl-N-(3-butenyl)amine 95%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
N-Benzylbut-3-en-1-amine
OR30936-1G
OR30936-250MG
APOSOR30936-1GEA
212
EUR
APOSOR30936-1G
APOSOR30936-250MG
N-Benzyl-N-(3-butenyl)amine 95%
N-Benzyl-N-(3-butenyl)amine
Formula:
C₁₁H₁₅N MW: 161,25 g/mol Boiling Pt: 65 °C/0,75mmHg Storage Temperature: Refrigerator |
MDL Number:
MFCD01249643 CAS Number: 17150-62-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...