4-Pyrrolidin-1-ylpyridin-2-amine

Supplier: Apollo Scientific
Warning

OR30944-1G OR30944-5G
APOSOR30944-1GEA 80 EUR
APOSOR30944-1G APOSOR30944-5G
4-Pyrrolidin-1-ylpyridin-2-amine
4-Pyrrolidin-1-ylpyridin-2-amine
Formula: C₉H₁₃N₃
Storage Temperature: Refrigerator
MDL Number: MFCD08062967
CAS Number: 722550-01-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR