4-Chloro-6-ethylpyrimidin-2-amine 98%

Supplier: Apollo Scientific
Danger

Synonyms: 2-Amino-4-chloro-6-ethylpyrimidine

OR30960-1G OR30960-5G
APOSOR30960-1GEA 115 EUR
APOSOR30960-1G APOSOR30960-5G
4-Chloro-6-ethylpyrimidin-2-amine 98%
4-Chloro-6-ethylpyrimidin-2-amine
Formula: C₆H₈ClN₃
MW: 157,6 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09800366
CAS Number: 5734-67-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR