Methyl-3-phenylpropionate 98%

Supplier: Apollo Scientific

Synonyms: Methyl dihydrocinnamate, 3-Phenylpropionic acid methyl ester, Methyl 3-phenylpropionate, Hydrocinnamic acid methyl ester

OR30978-500G
APOSOR30978-500GEA 109 EUR
APOSOR30978-500G
Methyl-3-phenylpropionate 98%
Methyl 3-phenylpropionate
Formula: C₁₀H₁₂O₂
MW: 164,2 g/mol
Boiling Pt: 236…239 °C
Density: 1,05 g/cm³ (25 °C)
Flash Pt: 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00017209
CAS Number: 103-25-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR