2-Bromostyrene 97+%
Supplier: Apollo Scientific
|
Warning
|
OR3099-5G
OR3099-100G
OR3099-25G
APOSOR3099-5GEA
36.4
EUR
APOSOR3099-5G
APOSOR3099-100G
APOSOR3099-25G
2-Bromostyrene 97+%
2-Bromostyrene
Formula:
C₈H₇Br MW: 183,05 g/mol Boiling Pt: 102…104 °C/22mmHg Melting Pt: –53 °C Density: 1,46 g/cm³ (25 °C) Flash Pt: 86 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00000076 CAS Number: 2039-88-5 |
Learn more
![](/m-ie.vwr.com/en_IE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...