1,4,5,6-Tetrahydrocyclopenta[c]pyrazole-3-carboxylic acid

Supplier: Apollo Scientific
Warning

Synonyms: 1H,4H,5H,6H-Cyclopenta[c]pyrazole-3-carboxylic acid

OR310021-1G OR310021-5G
APOSOR310021-1GEA 108 EUR
APOSOR310021-1G APOSOR310021-5G
1,4,5,6-Tetrahydrocyclopenta[c]pyrazole-3-carboxylic acid
1,4,5,6-Tetrahydrocyclopenta[c]pyrazole-3-carboxylic acid
Formula: C₇H₈N₂O₂
MW: 152,15 g/mol
Melting Pt: >250 °C
Storage Temperature: Ambient
MDL Number: MFCD01248822
CAS Number: 5932-32-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR