6-Chloropyridazine-3-carboxamide 97%

Supplier: Apollo Scientific
Warning

OR3101-1G OR3101-5G OR3101-2.5G OR3101-10G
APOSOR3101-1GEA 109 EUR
APOSOR3101-1G APOSOR3101-5G APOSOR3101-2.5G APOSOR3101-10G
6-Chloropyridazine-3-carboxamide 97%
6-Chloropyridazine-3-carboxamide
Formula: C₅H₄ClN₃O
MW: 157,5600 g/mol
Melting Pt: 243…244 °C
Storage Temperature: Ambient
MDL Number: MFCD09258882
CAS Number: 66346-83-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR