4-Methyl-3-thiopheneboronic acid

Supplier: Apollo Scientific

OR310105-1G OR310105-500MG
APOSOR310105-1GEA 364 EUR
APOSOR310105-1G APOSOR310105-500MG
4-Methyl-3-thiopheneboronic acid
4-Methyl-3-thiopheneboronic acid
Formula: C₅H₇BO₂S
MW: 141,99 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04039882
CAS Number: 177735-11-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR