1-Butylboronic acid-(1S,2S,3R,5S)-(+)-2,3-pinanediol ester

Supplier: Apollo Scientific

OR310106-1G OR310106-500MG
APOSOR310106-1GEA 364 EUR
APOSOR310106-1G APOSOR310106-500MG
1-Butylboronic acid-(1S,2S,3R,5S)-(+)-2,3-pinanediol ester
1-Butylboronic acid-(1S,2S,3R,5S)-(+)-2,3-pinanediol ester
Formula: C₁₄H₂₅BO₂
MW: 236,17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09037499
CAS Number: 85167-10-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR