3-Iodo-1H-indole-2-carbonitrile
Supplier: Apollo Scientific
OR310143-1G
OR310143-500MG
APOSOR310143-1GEA
487
EUR
APOSOR310143-1G
APOSOR310143-500MG
3-Iodo-1H-indole-2-carbonitrile
3-Iodo-1H-indole-2-carbonitrile
Formula:
C₉H₅IN₂ Melting Pt: 178…180 °C Storage Temperature: Ambient |
MDL Number:
MFCD22126121 CAS Number: 51796-65-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...