4-Bromobutylboronic acid (1S,2S,3R,5S)-(+)-2,3-pinanediol ester

Supplier: Apollo Scientific

OR310152-1G OR310152-500MG
APOSOR310152-1GEA 487 EUR
APOSOR310152-1G APOSOR310152-500MG
4-Bromobutylboronic acid (1S,2S,3R,5S)-(+)-2,3-pinanediol ester
4-Bromobutylboronic acid (1S,2S,3R,5S)-(+)-2,3-pinanediol ester
Formula: C₁₄H₂₄BBrO₂
MW: 315,06 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09037500
CAS Number: 165881-36-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR