4-Cyano-3-pyridineboronic acid pinacol ester

Supplier: Apollo Scientific

Synonyms: 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)isonicotinonitrile, 4-Cyanopyridine-3-boronic acid, pinacol ester, 4-Cyanopyridine-3-boronic acid pinacol ester

OR310221-1G OR310221-250MG OR310221-5G
APOSOR310221-1GEA 355 EUR
APOSOR310221-1G APOSOR310221-250MG APOSOR310221-5G
4-Cyano-3-pyridineboronic acid pinacol ester
4-Cyano-3-pyridineboronic acid pinacol ester
Formula: C₁₂H₁₅BN₂O₂
MW: 230,07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08458478
CAS Number: 878194-91-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR