2-Methoxypyrimidine-5-carboxylic acid

Supplier: Apollo Scientific

OR310777-1G OR310777-5G
APOSOR310777-1GEA 168 EUR
APOSOR310777-1G APOSOR310777-5G
2-Methoxypyrimidine-5-carboxylic acid
2-Methoxypyrimidine-5-carboxylic acid
Formula: C₆H₆N₂O₃
Storage Temperature: Ambient
MDL Number: MFCD08144700
CAS Number: 344325-95-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR