5-Chloro-1H-pyrrolo[2,3-c]pyridine
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
5-Chloro-6-azaindole
OR310792-1G
OR310792-5G
APOSOR310792-1GEA
64
EUR
APOSOR310792-1G
APOSOR310792-5G
5-Chloro-1H-pyrrolo[2,3-c]pyridine
5-Chloro-1H-pyrrolo[2,3-c]pyridine
Formula:
C₇H₅ClN₂ MW: 152,5800 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD09907859 CAS Number: 131084-55-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...