Methyl-2-amino-3-hydroxybenzoate 98%

Supplier: Apollo Scientific
Warning

Synonyms: Methyl 2-amino-3-hydroxybenzoate

OR310800-1G OR310800-250MG OR310800-5G
APOSOR310800-1GEA 119 EUR
APOSOR310800-1G APOSOR310800-250MG APOSOR310800-5G
Methyl-2-amino-3-hydroxybenzoate 98%
Methyl-2-amino-3-hydroxybenzoate
Formula: C₈H₉NO₃
MW: 167,17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01675233
CAS Number: 17672-21-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR