Methyl-3-hydroxy-4-methylbenzoate 98%

Supplier: Apollo Scientific

Synonyms: Methyl 3-hydroxy-4-methylbenzoate

OR311176-25G OR311176-5G
APOSOR311176-25GEA 437 EUR
APOSOR311176-25G APOSOR311176-5G
Methyl-3-hydroxy-4-methylbenzoate 98%
Methyl-3-hydroxy-4-methylbenzoate
Formula: C₉H₁₀O₃
MW: 166,18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00213808
CAS Number: 3556-86-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR