Dimethyl-N-(tert-butoxycarbonyl)-L-glutamate 96%

Supplier: Apollo Scientific
Warning

Synonyms: N-Boc-L-glutamic Acid Dimethyl Ester, N-Boc-L-glutamic acid 1,5-dimethyl ester, Dimethyl N-Boc-L-glutamate, (S)-Dimethyl 2-((tert-butoxycarbonyl)amino)pentanedioate, N-(tert-Butoxycarbonyl)-L-glutamic Acid Dimethyl Ester, Boc-Glu(OMe)-OMe

OR313088-100G OR313088-25G OR313088-5G
APOSOR313088-100GEA 241 EUR
APOSOR313088-100G APOSOR313088-25G APOSOR313088-5G
Dimethyl-N-(tert-butoxycarbonyl)-L-glutamate 96%
Dimethyl-N-(tert-butoxycarbonyl)-L-glutamate
Formula: C₁₂H₂₁NO₆
MW: 275,30 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03094774
CAS Number: 59279-60-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR