9-Bromo-1-nonanol 97%

Supplier: Apollo Scientific
Warning

OR3134-25G OR3134-100G OR3134-500G
APOSOR3134-25GEA 66.7 EUR
APOSOR3134-25G APOSOR3134-100G APOSOR3134-500G
9-Bromo-1-nonanol 97%
9-Bromo-1-nonanol
Formula: C₉H₁₉BrO
Boiling Pt: 125…126 °C/2mmHg
Melting Pt: 33…35 °C
Flash Pt: 109 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00063348
CAS Number: 55362-80-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR