Pyridine-3,5-dicarboxylic acid 98%

Supplier: Apollo Scientific
Warning

Synonyms: Dinicotinic acid, 3,5-Pyridinedicarboxylic acid

OR315024-100G OR315024-25G OR315024-500G
APOSOR315024-100GEA 120 EUR
APOSOR315024-100G APOSOR315024-25G APOSOR315024-500G
Pyridine-3,5-dicarboxylic acid 98%
Pyridine-3,5-dicarboxylic acid
Formula: C₇H₅NO₄
MW: 167.12 g/mol
Boiling Pt: 357
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00006393
CAS Number: 499-81-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR