2,7-Di-tert-butylfluorene
Supplier: Apollo Scientific
OR315036-25G
OR315036-5G
APOSOR315036-25GEA
61.1
EUR
APOSOR315036-25G
APOSOR315036-5G
2,7-Di-tert-butylfluorene
2,7-Di-tert-butylfluorene
Formula:
C₂₁H₂₆ MW: 278,43 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03093998 CAS Number: 58775-05-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...