3-Amino-2-methoxy-6-picoline 98%
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
3-Amino-2-methoxy-6-methylpyridine
OR315073-1G
OR315073-5G
OR315073-25G
APOSOR315073-1GEA
78.6
EUR
APOSOR315073-1G
APOSOR315073-5G
APOSOR315073-25G
3-Amino-2-methoxy-6-picoline 98%
3-Amino-2-methoxy-6-picoline
Formula:
C₇H₁₀N₂O Storage Temperature: Ambient |
MDL Number:
MFCD03095079 CAS Number: 186413-79-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...