5-(Hydroxymethyl)furan-2-carboxylic Acid 95%

Supplier: Apollo Scientific
Warning

Synonyms: 5-Hydroxymethyl-2-furancarboxylic acid, Sumiki's acid, 5-(Hydroxymethyl)-2-furoic acid

OR315124-1G OR315124-5G OR315124-25G
APOSOR315124-1GEA 31.9 EUR
APOSOR315124-1G APOSOR315124-5G APOSOR315124-25G
5-(Hydroxymethyl)furan-2-carboxylic Acid 95%
5-(Hydroxymethyl)furan-2-carboxylic Acid
Formula: C6H6O4
Melting Pt: 165 °C
Storage Temperature: Freezer
MDL Number: MFCD03274472
CAS Number: 6338-41-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR