H-Aib-OtBu·HCl 97%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
tert-Butyl 2-aminoisobutyrate hydrochloride, tert-Butyl 2-amino-2-methylpropanoate hydrochloride
OR315173-25G
OR315173-5G
OR315173-100G
APOSOR315173-25GEA
200
EUR
APOSOR315173-25G
APOSOR315173-5G
APOSOR315173-100G
H-Aib-OtBu·HCl 97%
H-Aib-OtBu·HCl
Formula:
C₈H₁₇NO₂·HCl MW: 195,7 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD08272284 CAS Number: 84758-81-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...