8-Benzyl-2,8-diazaspiro[4.5]decan-1-one 95+%

Supplier: Apollo Scientific

OR315196-10G OR315196-1G OR315196-5G
APOSOR315196-10GEA 2770 EUR
APOSOR315196-10G APOSOR315196-1G APOSOR315196-5G
8-Benzyl-2,8-diazaspiro[4.5]decan-1-one 95+%
8-Benzyl-2,8-diazaspiro[4.5]decan-1-one
Formula: C₁₅H₂₀N₂O
Storage Temperature: Ambient
MDL Number: MFCD11109549
CAS Number: 1123242-53-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR