4-Chloro-2-methoxypyridine

Supplier: Apollo Scientific
Warning

Synonyms: 2-Methoxy-4-chloropyridine

OR315215-1G OR315215-5G
APOSOR315215-1GEA 58.2 EUR
APOSOR315215-1G APOSOR315215-5G
4-Chloro-2-methoxypyridine
4-Chloro-2-methoxypyridine
Formula: C₆H₆ClNO
MW: 143,5709 g/mol
Melting Pt: 26 °C
Storage Temperature: Ambient
MDL Number: MFCD06738656
CAS Number: 72141-44-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR