5-Iodo-2-methoxypyridine 98%

Supplier: Apollo Scientific

OR315217-10G OR315217-5G
APOSOR315217-10GEA 90.2 EUR
APOSOR315217-10G APOSOR315217-5G
5-Iodo-2-methoxypyridine 98%
5-Iodo-2-methoxypyridine
Formula: C₆H₆INO
Storage Temperature: Ambient
MDL Number: MFCD07781180
CAS Number: 13472-61-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR