2-Butynoic acid 99%

Supplier: Apollo Scientific
Danger

OR315221-25G OR315221-5G
APOSOR315221-25GEA 65.5 EUR
APOSOR315221-25G APOSOR315221-5G
2-Butynoic acid 99%
2-Butynoic acid
Formula: C₄H₄O₂
MW: 84,07 g/mol
Boiling Pt: 200…203 °C
Melting Pt: 78…80 °C
Density: 0,964 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004363
CAS Number: 590-93-2
EINECS: 209-695-7
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR