3-Iodophenyl isothiocyanate 98%
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
3-Iodophenylisothiocyanate, 1-Iodo-3-isothiocyanatobenzene
OR315243-1G
OR315243-25G
OR315243-5G
APOSOR315243-1GEA
21.9
EUR
APOSOR315243-1G
APOSOR315243-25G
APOSOR315243-5G
3-Iodophenyl isothiocyanate 98%
3-Iodophenyl isothiocyanate
Formula:
C₇H₄INS MW: 261,09 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00041074 CAS Number: 3125-73-3 UN: 2811 ADR: 6.1,III |
Learn more
![](/m-ie.vwr.com/en_IE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...