5-Norbornene-2-methanol 99%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
Bicyclo[2.2.1]hept-5-ene-2-methanol, 5-Norbornen-2-methanol (mixture of isomers), 2-Hydroxymethyl-5-norbornene, Bicyclo[2.2.1]hept-5-en-2-ylmethanol
OR315262-25G
OR315262-100G
OR315262-5G
APOSOR315262-25GEA
21.9
EUR
APOSOR315262-25G
APOSOR315262-100G
APOSOR315262-5G
5-Norbornene-2-methanol 99%
5-Norbornene-2-methanol
Formula:
C₈H₁₂O MW: 124,183 g/mol Boiling Pt: 120 °C Density: 1,03 g/cm³ (25 °C) Flash Pt: 87 °C Storage Temperature: Ambient |
MDL Number:
MFCD00167571 CAS Number: 95-12-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...