(S)-Oxiran-2-ylmethyl-3-nitrobenzenesulphonate 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
(S)-(+)-Glycidyl nosylate, (S)-Oxiran-2-ylmethyl 3-nitrobenzenesulphonate, Glycidyl (S)-(+)-3-nitrobenzenesulphonate
OR315284-25G
OR315284-100G
APOSOR315284-25GEA
77.1
EUR
APOSOR315284-25G
APOSOR315284-100G
(S)-Oxiran-2-ylmethyl-3-nitrobenzenesulphonate 98%
(S)-Oxiran-2-ylmethyl-3-nitrobenzenesulphonate
Formula:
C₉H₉NO₆S MW: 259,23 g/mol Melting Pt: 63…66 °C Storage Temperature: Ambient |
MDL Number:
MFCD00064582 CAS Number: 115314-14-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...