(S)-(-)-1-Phenylethanol 99%

Supplier: Apollo Scientific
Danger

OR315315-100G OR315315-25G OR315315-500G
APOSOR315315-100GEA 71.2 EUR
APOSOR315315-100G APOSOR315315-25G APOSOR315315-500G
(S)-(-)-1-Phenylethanol 99%
(S)-(-)-1-Phenylethanol
Formula: C₈H₁₀O
Boiling Pt: 98 °C/20,25mmHg
Density: 0,996 g/cm³ (25 °C)
Flash Pt: 86 °C
Storage Temperature: Ambient
MDL Number: MFCD00064264
CAS Number: 1445-91-6
UN: 2937
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR