1,2,3,4-Tetrahydrobenzo[b]azepin-5-one 96%

Supplier: Apollo Scientific

Synonyms: 1,2,3,4-Tetrahydro-benzo[b]azepin-5-one, 1,2,3,4-Tetrahydrobenzazepine-5-one, 2,3,4,5-tetrahydro-1H-1-benzazepin-5-one

OR315598-10G OR315598-5G
APOSOR315598-10GEA 111 EUR
APOSOR315598-10G APOSOR315598-5G
1,2,3,4-Tetrahydrobenzo[b]azepin-5-one 96%
1,2,3,4-Tetrahydrobenzo[b]azepin-5-one
Formula: C₁₀H₁₁NO
MW: 161,204 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03426404
CAS Number: 1127-74-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR