Benzyltrimethylammonium dichloroiodate 97%

Supplier: Apollo Scientific
Warning

OR316020-100G OR316020-25G OR316020-5G
APOSOR316020-100GEA 183 EUR
APOSOR316020-100G APOSOR316020-25G APOSOR316020-5G
Benzyltrimethylammonium dichloroiodate 97%
Benzyltrimethylammonium dichloroiodate
Formula: C₁₀H₁₆Cl₂IN
Melting Pt: 124…126 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00075259
CAS Number: 114971-52-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR