4-Bromo-3-methylbenzamide

Supplier: Apollo Scientific
Warning

OR3168-25G
APOSOR3168-25GEA 91.5 EUR
APOSOR3168-25G
4-Bromo-3-methylbenzamide
4-Bromo-3-methylbenzamide
Formula: C₈H₈BrNO
MW: 214,06 g/mol
Melting Pt: 177…182 °C
Storage Temperature: Ambient
MDL Number: MFCD00672914
CAS Number: 170229-98-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR