Ethyl-1-boc-3-azetidineacetate
Supplier: Apollo Scientific
Synonyms:
3-(2-Ethoxy-2-oxoethyl)azetidine-1-carboxylic acid tert-Butyl ester, Ethyl 1-(tert-Butoxycarbonyl)-3-azetidineacetate, 1-(tert-Butoxycarbonyl)-3-(2-ethoxy-2-oxoethyl)azetidine, 1-Boc-3-(2-ethoxy-2-oxoethyl)azetidine, tert-Butyl 3-(2-Ethoxy-2-oxoethyl)azetidine-1-carboxylate, Ethyl 1-Boc-3-azetidineacetate, 1-(tert-Butoxycarbonyl)-3-azetidineacetic acid ethyl ester
OR317235-1G
OR317235-5G
APOSOR317235-1GEA
80
EUR
APOSOR317235-1G
APOSOR317235-5G
Ethyl-1-boc-3-azetidineacetate
Ethyl-1-boc-3-azetidineacetate
Formula:
C₁₂H₂₁NO₄ MW: 243,31 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11616049 CAS Number: 158602-35-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...