(R)-Azetidine-2-carboxylic acid

Supplier: Apollo Scientific

OR317238-10G OR317238-1G OR317238-5G
APOSOR317238-10GEA 464 EUR
APOSOR317238-10G APOSOR317238-1G APOSOR317238-5G
(R)-Azetidine-2-carboxylic acid
(R)-Azetidine-2-carboxylic acid
Formula: C₄H₇NO₂
MW: 101,1039 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00800731
CAS Number: 7729-30-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR