1,2,4-Triazole-3-carboxylic acid 95%

Supplier: Apollo Scientific
Warning

Synonyms: 1H-1,2,4-Triazole-3-carboxylic acid

OR3176-1G OR3176-25G OR3176-5G
APOSOR3176-1GEA 21.9 EUR
APOSOR3176-1G APOSOR3176-25G APOSOR3176-5G
1,2,4-Triazole-3-carboxylic acid 95%
1,2,4-Triazole-3-carboxylic acid
Formula: C₃H₃N₃O₂
MW: 113,0748 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00078268
CAS Number: 4928-87-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR