Imidazo[1,2-a]pyridine-2-carbaldehyde 95+%

Supplier: Apollo Scientific

Synonyms: Imidazo[1,2-a]pyridine-2-carboxaldehyde

OR318013-1G OR318013-5G OR318013-10G
APOSOR318013-1GEA 410 EUR
APOSOR318013-1G APOSOR318013-5G APOSOR318013-10G
Imidazo[1,2-a]pyridine-2-carbaldehyde 95+%
Imidazo[1,2-a]pyridine-2-carbaldehyde
Formula: C₈H₆N₂O
MW: 146,1460 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06739247
CAS Number: 118000-43-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR