3-(3,5-Bis-methanesulphonylphenyl)propionic acid 95+%

Supplier: Apollo Scientific

OR318027-1G OR318027-5G
APOSOR318027-1GEA 318 EUR
APOSOR318027-1G APOSOR318027-5G
3-(3,5-Bis-methanesulphonylphenyl)propionic acid 95+%
3-(3,5-Bis-methanesulphonylphenyl)propionic acid
Formula: C₁₁H₁₄O₆S₂
MW: 306,36 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10688275
CAS Number: 1040682-15-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR