Ethyl-4-ethoxy-3-nitrobenzoate 95+%

Supplier: Apollo Scientific

OR318054-1G OR318054-10G OR318054-5G
APOSOR318054-1GEA 159 EUR
APOSOR318054-1G APOSOR318054-10G APOSOR318054-5G
Ethyl-4-ethoxy-3-nitrobenzoate 95+%
Ethyl-4-ethoxy-3-nitrobenzoate
Formula: C₁₁H₁₃NO₅
MW: 239,23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08692632
CAS Number: 937625-32-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR