5-(5-Bromo-2-thienyl)-1H-pyrazole

Supplier: Apollo Scientific
Warning

Synonyms: 5-(5-Bromothiophen-2-yl)-1H-pyrazole

OR32-1G OR32-100MG OR32-250MG
APOSOR32-1GEA 150 EUR
APOSOR32-1G APOSOR32-100MG APOSOR32-250MG
5-(5-Bromo-2-thienyl)-1H-pyrazole
5-(5-Bromo-2-thienyl)-1H-pyrazole
Formula: C₇H₅BrN₂S
MW: 229,1 g/mol
Melting Pt: 167 °C
Storage Temperature: Ambient
MDL Number: MFCD00120516
CAS Number: 166196-67-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR