3-Bromo-5-methylpyridine

Supplier: Apollo Scientific
Danger

OR3203-5G OR3203-1G OR3203-10G
APOSOR3203-5GEA 33.5 EUR
APOSOR3203-5G APOSOR3203-1G APOSOR3203-10G
3-Bromo-5-methylpyridine
3-Bromo-5-methylpyridine
Formula: C₆H₆BrN
Density: 1,487 g/cm³ (25 °C)
Flash Pt: 90 °C
Storage Temperature: Ambient
MDL Number: MFCD01646141
CAS Number: 3430-16-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR