2-Chloro-6-methoxybenzoic acid

Supplier: Apollo Scientific

OR321002-1G OR321002-5G
APOSOR321002-1GEA 66.9 EUR
APOSOR321002-1G APOSOR321002-5G
2-Chloro-6-methoxybenzoic acid
2-Chloro-6-methoxybenzoic acid
Formula: C₈H₇ClO₃
Storage Temperature: Ambient
MDL Number: MFCD00127702
CAS Number: 3260-89-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR